Stampa…

2-Bromo-4-(trifluoromethoxy)aniline ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 2-Bromo-4-trifluoromethoxyaniline

B3799-5G B3799-25G
TCIAB3799-5GEA 38 EUR
TCIAB3799-5G TCIAB3799-25G
2-Bromo-4-(trifluoromethoxy)aniline ≥98.0% (by GC)
2-Bromo-4-(trifluoromethoxy)aniline
Formula: C₇H₅BrF₃NO
MDL Number: MFCD00203478
CAS Number: 175278-17-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12563bb820920f02aac1fed8716c9fde9cb9d757&vid=12563bb820920f02aac1fed8716c9fde9cb9d757&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR