Stampa…

trans-4-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: p-Coumaric acid, trans-p-Cumaric Acid, trans-p-Coumaric Acid

C0393-25G C0393-500G C0393-100G
TCIAC0393-25GEA 35 EUR
TCIAC0393-25G TCIAC0393-500G TCIAC0393-100G
trans-4-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-4-Hydroxycinnamic acid
Formula: C₉H₈O₃
Punto di fusione: 214°C
MDL Number: MFCD00004399
CAS Number: 501-98-4
EINECS: 231-000-0
Merck Index: 14,02560
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5876702a5034ac1ddae38171322c8bb1217a2c02&vid=5876702a5034ac1ddae38171322c8bb1217a2c02&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR