Stampa…

3-Bromo-5-methoxypyridine ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 5-Bromo-3-methoxypyridine

B3684-5G B3684-1G
TCIAB3684-5GEA 110 EUR
TCIAB3684-5G TCIAB3684-1G
3-Bromo-5-methoxypyridine ≥97.0% (by titrimetric analysis)
3-Bromo-5-methoxypyridine
Formula: C₆H₆BrNO
MDL Number: MFCD00234169
CAS Number: 50720-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d96cdc5a3eb516fd38d8b488321ceb97e8c35a&vid=b0d96cdc5a3eb516fd38d8b488321ceb97e8c35a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR