Stampa…

3,4-Dimethoxybenzoyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Veratroyl chloride, 3,4-Dimethoxybenzene-1-carbonyl chloride

D4093-5G D4093-25G
TCIAD4093-5GEA 56 EUR
TCIAD4093-5G TCIAD4093-25G
3,4-Dimethoxybenzoyl chloride ≥98.0% (by GC, titration analysis)
3,4-Dimethoxybenzoyl chloride
Formula: C₉H₉ClO₃
Peso molecolare: 200,62 g/mol
Punto di ebollizione: 95 - 98 °C (1 -rr)
Punto di fusione: 71 °C
MDL Number: MFCD00000674
CAS Number: 3535-37-3
EINECS: 222-568-0
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15930e268e3d2ef7ff0200a654c60262b857b938&vid=15930e268e3d2ef7ff0200a654c60262b857b938&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR