2-Bromo-4,5-difluorophenol ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₃BrF₂O Flash Pt: 81°C |
MDL Number:
MFCD00070751 CAS Number: 166281-37-4 |
|
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...