Stampa…

2-Fluoro-p-toluic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

F0813-5G F0813-1G
TCIAF0813-5GEA 160 EUR
TCIAF0813-5G TCIAF0813-1G
2-Fluoro-p-toluic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-p-toluic acid
Formula: C₈H₇FO₂
MDL Number: MFCD03092906
CAS Number: 7697-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=771973e500ae2d110f14a4d503e3241dfc5e4cba&vid=771973e500ae2d110f14a4d503e3241dfc5e4cba&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR