Stampa…

Trimetilamina ∼25% (w/w) ∼3.2 mol/L in methanol

Fornitore: TCI EUROPE
Pericolo

T2268-500ML T2268-100ML
TCIAT2268-500MLEA 63 EUR
TCIAT2268-500ML TCIAT2268-100ML
Trimetilamina ∼25% (w/w) ∼3.2 mol/L in methanol
Trimetilamina
Formula: C₃H₉N
Peso molecolare: 59,11 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 2924
ADR: 3,II
Merck Index: 14,09710
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab7ef2cc7e2b7c936ce95e216ff7a4425ec52147&vid=88c044e92baed75b10440243f55fdfa15a97e641&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless clear liquid
Concentration (Neutralization titration) 25.0 to 27.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR