2-Bromo-3-methoxypyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B3861-5G
B3861-25G
TCIAB3861-5GEA
79
EUR
TCIAB3861-5G
TCIAB3861-25G
2-Bromo-3-methoxypyridine ≥98.0% (by GC)
2-Bromo-3-methoxypyridine
Formula:
C₆H₆BrNO Peso molecolare: 188,02 g/mol |
MDL Number:
MFCD01570896 CAS Number: 24100-18-3 EINECS: 246-017-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...