Stampa…

2-Bromo-3-chloropyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B3860-5G B3860-25G
TCIAB3860-5GEA 51 EUR
TCIAB3860-5G TCIAB3860-25G
2-Bromo-3-chloropyridine ≥98.0% (by GC)
2-Bromo-3-chloropyridine
Formula: C₅H₃BrClN
MDL Number: MFCD04039344
CAS Number: 96424-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c470ed556038116ee9fae861b812922b0d7efa1d&vid=c470ed556038116ee9fae861b812922b0d7efa1d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR