2',4'-Dichlorovalerophenone ≥95.0% (by GC)
Fornitore: TCI
Formula:
C₁₁H₁₂Cl₂O |
MDL Number:
MFCD09028061 CAS Number: 61023-66-3 |
|
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...