Stampa…

α,α,α,3,4-Pentachlorotoluene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3,4-Dichlorobenzotrichloride, 1,2-Dichloro-4-(trichloromethyl)benzene

D2481-25G D2481-500G
TCIAD2481-25GEA 28 EUR
TCIAD2481-25G TCIAD2481-500G
α,α,α,3,4-Pentachlorotoluene ≥98.0% (by GC)
α,α,α,3,4-Pentachlorotoluene
Formula: C₇H₃Cl₅
Peso molecolare: 264,3637 g/mol
MDL Number: MFCD00018820
CAS Number: 13014-24-9
EINECS: 235-869-7
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4d2cf00d54ecfd7180c506b51e23c2059bd056c&vid=e4d2cf00d54ecfd7180c506b51e23c2059bd056c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR