Stampa…

4-Phenoxyphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid

P1974-25G P1974-5G
TCIAP1974-25GEA 248 EUR
TCIAP1974-25G TCIAP1974-5G
4-Phenoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Phenoxyphenylboronic acid
Formula: C₁₂H₁₁BO₃
Peso molecolare: 214,03 g/mol
Punto di fusione: 115°C
MDL Number: MFCD00093312
CAS Number: 51067-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43e97b28e7800b18a4ae69ad884782cb3005cd02&vid=43e97b28e7800b18a4ae69ad884782cb3005cd02&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR