Stampa…

4-Amino-3-chlorobenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

A1962-25G A1962-5G
TCIAA1962-25GEA 194 EUR
TCIAA1962-25G TCIAA1962-5G
4-Amino-3-chlorobenzonitrile ≥98.0% (by GC)
4-Amino-3-chlorobenzonitrile
Formula: C₇H₅ClN₂
Peso molecolare: 152,58 g/mol
MDL Number: MFCD00052917
CAS Number: 21803-75-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f801ef1613a3637bc5fefcdd9f2a036bd04b3af4&vid=f801ef1613a3637bc5fefcdd9f2a036bd04b3af4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR