Stampa…

1,1,1,3,3,3-Hexafluoroisopropyl methacrylate ≥98.0% (by GC) stabilizzato

Fornitore: TCI EUROPE
Pericolo

Synonyms: Hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-Hexafluoroisopropylmethacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M0988-10G M0988-25G M0988-5G
TCIAM0988-10GEA 66 EUR
TCIAM0988-10G TCIAM0988-25G TCIAM0988-5G
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate ≥98.0% (by GC) stabilizzato
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate
Formula: C₇H₆F₆O₂
Peso molecolare: 236,12 g/mol
Flash Pt: 16°C
MDL Number: MFCD00040105
CAS Number: 3063-94-3
EINECS: 221-309-9
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb1bbf978c7ffa28b2cc3db89f4b0db2b84e8443&vid=23cbef8f591a88d6ae3436301e3e0f9e063f68e7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR