Stampa…

2-Amino-4'-chlorobenzophenone ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (2-Aminophenyl)(4-chlorophenyl)methanone

A1244-25G A1244-5G
TCIAA1244-25GEA 162 EUR
TCIAA1244-25G TCIAA1244-5G
2-Amino-4'-chlorobenzophenone ≥98.0% (by GC, titration analysis)
2-Amino-4'-chlorobenzophenone
Formula: C₁₃H₁₀ClNO
Peso molecolare: 231,6800 g/mol
MDL Number: MFCD00025193
CAS Number: 2894-51-1
EINECS: 220-770-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd405e20af9cef2e44ddf439e268895eb6888861&vid=dd405e20af9cef2e44ddf439e268895eb6888861&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 101.0 to 105.0 deg-C
Solubility in Chloroform almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR