3-Methylhippuric acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₁₀H₁₁NO₃ |
MDL Number:
MFCD00044399 CAS Number: 27115-49-7 |
|
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...