Stampa…

6-Amino-2,4-xylenol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-4,6-dimethylphenol

A0962-25G A0962-5G
TCIAA0962-25GEA 405 EUR
TCIAA0962-25G TCIAA0962-5G
6-Amino-2,4-xylenol ≥98.0% (by GC, titration analysis)
6-Amino-2,4-xylenol
Formula: C₈H₁₁NO
Peso molecolare: 137,1790 g/mol
MDL Number: MFCD00007691
CAS Number: 41458-65-5
EINECS: 255-380-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0c9a186d1ed171b068c07da84fef15a4fe9c3c&vid=5f0c9a186d1ed171b068c07da84fef15a4fe9c3c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 130.0 to 136.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR