2,4,6-T (2,4,6-trichlorophenoxy acetic acid) ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-(2,4,6-Trichlorophenoxy)acetic acid
T1063-25G
T1063-5G
TCIAT1063-25GEA
373
EUR
TCIAT1063-25G
TCIAT1063-5G
2,4,6-T (2,4,6-trichlorophenoxy acetic acid) ≥98.0% (by GC, titration analysis)
2,4,6-T (2,4,6-trichlorophenoxy acetic acid)
Formula:
C₈H₅Cl₃O₃ Peso molecolare: 255,4825 g/mol |
MDL Number:
MFCD00053498 CAS Number: 575-89-3 EINECS: 209-394-0 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...