Stampa…

Hexyl chloroformate ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: Hexyl carbonochloridate

C0723-25ML C0723-5ML
TCIAC0723-25MLEA 203 EUR
TCIAC0723-25ML TCIAC0723-5ML
Hexyl chloroformate ≥98.0% (by GC, titration analysis)
Hexyl chloroformate
Formula: C₇H₁₃ClO₂
Peso molecolare: 164.6299 g/mol
Punto di ebollizione: 78°C
Flash Pt: 61°C
MDL Number: MFCD00000651
CAS Number: 6092-54-2
EINECS: 228-036-4
UN: 3277
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1654c4a92df034055256cf0b69edb4d16232a3&vid=af1654c4a92df034055256cf0b69edb4d16232a3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR