Stampa…

Heptyl chloroformate ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: Heptyl carbonochloridate

C0722-25ML C0722-5ML
TCIAC0722-25MLEA 155 EUR
TCIAC0722-25ML TCIAC0722-5ML
Heptyl chloroformate ≥98.0% (by GC, titration analysis)
Heptyl chloroformate
Formula: C₈H₁₅ClO₂
Punto di ebollizione: 71°C
MDL Number: MFCD00059484
CAS Number: 33758-34-8
UN: 3277
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8890004e4dbc9b5afdb7eb9286173bd0b04c0e25&vid=8890004e4dbc9b5afdb7eb9286173bd0b04c0e25&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR