3-Ethynyltoluene ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
1-Ethynyl-3-methylbenzene, m-Tolylacetylene
Formula:
C₉H₈ Punto di ebollizione: 60°C Flash Pt: 25°C |
MDL Number:
MFCD01318174 CAS Number: 766-82-5 UN: 3295 ADR: 3,III |
|
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...