Ethyl pentafluorobenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Ethyl 2,3,4,5,6-pentafluorobenzoate
P1419-25G
P1419-5G
TCIAP1419-25GEA
56
EUR
TCIAP1419-25G
TCIAP1419-5G
Ethyl pentafluorobenzoate ≥98.0% (by GC)
Ethyl pentafluorobenzoate
Formula:
C₉H₅F₅O₂ Peso molecolare: 240,13 g/mol Flash Pt: 81°C |
MDL Number:
MFCD00039211 CAS Number: 4522-93-4 EINECS: 224-854-0 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...