2,2,2-Trifluoroethyl methanesulphonate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
M1193-25G
M1193-5G
TCIAM1193-25GEA
75
EUR
TCIAM1193-25G
TCIAM1193-5G
2,2,2-Trifluoroethyl methanesulphonate ≥98.0% (by GC)
2,2,2-Trifluoroethyl methanesulphonate
Formula:
C₃H₅F₃O₃S Peso molecolare: 178,130 g/mol Punto di ebollizione: 99°C Flash Pt: 104°C |
MDL Number:
MFCD00025058 CAS Number: 25236-64-0 UN: 3265 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...