2,2,3,3,4,4,4-Heptafluorobutylamine ≥95.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
H1300-1G
H1300-5G
TCIAH1300-1GEA
31
EUR
TCIAH1300-1G
TCIAH1300-5G
2,2,3,3,4,4,4-Heptafluorobutylamine ≥95.0% (by titrimetric analysis)
2,2,3,3,4,4,4-Heptafluorobutylamine
Formula:
C₄H₄F₇N Peso molecolare: 199,07 g/mol Punto di ebollizione: 69°C |
MDL Number:
MFCD00014817 CAS Number: 374-99-2 EINECS: 206-780-0 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 92.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...