Stampa…

Methyl pentafluoropropionate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Methyl 2,2,3,3,3-pentafluoropropanoate

P1352-25G P1352-5G
TCIAP1352-25GEA 82 EUR
TCIAP1352-25G TCIAP1352-5G
Methyl pentafluoropropionate ≥98.0% (by GC)
Methyl pentafluoropropionate
Formula: C₄H₃F₅O₂
Peso molecolare: 178,06 g/mol
Punto di ebollizione: 60°C
Flash Pt: 6°C
MDL Number: MFCD00000430
CAS Number: 378-75-6
EINECS: 206-828-0
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1585e1f1d3d599f084708819d25cabe06a89a65&vid=f1585e1f1d3d599f084708819d25cabe06a89a65&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR