2,2,2-Trifluoroethyl butyrate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Butyric acid 2,2,2-Trifluoroethyl ester
B2144-25G
B2144-5G
TCIAB2144-25GEA
205
EUR
TCIAB2144-25G
TCIAB2144-5G
2,2,2-Trifluoroethyl butyrate ≥98.0% (by GC)
2,2,2-Trifluoroethyl butyrate
Formula:
C₆H₉F₃O₂ Peso molecolare: 170,1297 g/mol Flash Pt: 24°C |
MDL Number:
MFCD00077609 CAS Number: 371-27-7 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...