Stampa…

DemNA (Decanoyl m-nitroaniline)

Fornitore: Cayman Chemical

90349-10 90349-5 90349-50 90349-25
CAYM90349-10EA 209 EUR
CAYM90349-10 CAYM90349-5 CAYM90349-50 CAYM90349-25
DemNA (Decanoyl m-nitroaniline)
DemNA (decanoyl-m-nitroaniline)
Formula: C₁₆H₂₄N₂O₃
CAS Number: 72298-61-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR