Stampa…

D-(+)-Cellobiose ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, β-Cellobiose, Cellose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

C0056-25G C0056-5G
TCIAC0056-25GEA 54 EUR
TCIAC0056-25G TCIAC0056-5G
D-(+)-Cellobiose ≥98.0% (by HPLC)
D-(+)-Cellobiose
Formula: C₁₂H₂₂O₁₁
Peso molecolare: 342,30 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00136034
CAS Number: 528-50-7
EINECS: 208-436-5
Merck Index: 14,01961
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26f388cce840a6b6642ab01a4ee23989d830657c&vid=26f388cce840a6b6642ab01a4ee23989d830657c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Specific rotation [a]20/D +32.0 to +35.0 deg (C=0.8, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR