Stampa…

Erucic acid ≥85.0% (by GC)

Fornitore: TCI
Attenzione

D0965-25G D0965-400G D0965-100G
TCIAD0965-25GEA 18 EUR
TCIAD0965-25G TCIAD0965-400G TCIAD0965-100G
Erucic acid ≥85.0% (by GC)
Erucic acid
Formula: C₂₂H₄₂O₂
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00063188
CAS Number: 112-86-7
EINECS: 204-011-3
Merck Index: 14,03674
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63706fb64340fd7d2aa398a2dd5aee96b4fd16a4&vid=63706fb64340fd7d2aa398a2dd5aee96b4fd16a4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to lump
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR