2-Bromo-4-tert-butylphenol ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
B3763-5G
B3763-25G
TCIAB3763-5GEA
51
EUR
TCIAB3763-5G
TCIAB3763-25G
2-Bromo-4-tert-butylphenol ≥97.0% (by GC, titration analysis)
2-Bromo-4-tert-butylphenol
Formula:
C₁₀H₁₃BrO Peso molecolare: 229,1100 g/mol Punto di ebollizione: 113°C Punto di fusione: 51°C |
MDL Number:
MFCD02682891 CAS Number: 2198-66-5 EINECS: 218-602-9 |
Risultati del test di specificazione
Appearance | White or Colorless to Yellow to Green powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...