Stampa…

N-Allylaniline ≥99.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

A0880-25ML A0880-500ML A0880-100ML
TCIAA0880-25MLEA 67 EUR
TCIAA0880-25ML TCIAA0880-500ML TCIAA0880-100ML
N-Allylaniline ≥99.0% (by GC)
N-Allylaniline
Formula: C₉H₁₁N
Peso molecolare: 133,19 g/mol
Flash Pt: 89°C
MDL Number: MFCD00008638
CAS Number: 589-09-3
EINECS: 209-633-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=449892752bdafa85fbc95f14ac73a295f3f4b45b&vid=449892752bdafa85fbc95f14ac73a295f3f4b45b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR