Stampa…

Chlorodimethyloctylsilane ≥96.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

D1827-100ML D1827-25ML
TCIAD1827-100MLEA 207 EUR
TCIAD1827-100ML TCIAD1827-25ML
Chlorodimethyloctylsilane ≥96.0% (by GC)
Chlorodimethyloctylsilane
Formula: C₁₀H₂₃ClSi
Peso molecolare: 206,83 g/mol
Punto di ebollizione: 220°C
Flash Pt: 92°C
MDL Number: MFCD00000511
CAS Number: 18162-84-0
EINECS: 242-044-5
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae4f658a736087ce26efb30f1ae88eb447d5488a&vid=ae4f658a736087ce26efb30f1ae88eb447d5488a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR