Stampa…

Di(2-pyridyl) ketone oxime ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2,2'-Dipyridyl ketoxime

D1709-1G D1709-5G
TCIAD1709-1GEA 49 EUR
TCIAD1709-1G TCIAD1709-5G
Di(2-pyridyl) ketone oxime ≥98.0% (by HPLC, titration analysis)
Di(2-pyridyl) ketone oxime
Formula: C₁₁H₉N₃O
Peso molecolare: 199.21 g/mol
MDL Number: MFCD00006311
CAS Number: 1562-95-4
EINECS: 216-349-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0ae5e039d70e22f8a08ec01d93e7c7cd9d8cb68&vid=f0ae5e039d70e22f8a08ec01d93e7c7cd9d8cb68&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR