2,3,3-Trimethylindolenine ≥97.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
|
Attenzione
|
T0766-25ML
T0766-500ML
T0766-100ML
TCIAT0766-25MLEA
38
EUR
TCIAT0766-25ML
TCIAT0766-500ML
TCIAT0766-100ML
2,3,3-Trimethylindolenine ≥97.0% (by total nitrogen basis)
2,3,3-Trimethylindolenine
Formula:
C₁₁H₁₃N Peso molecolare: 159,23 g/mol Punto di ebollizione: 229°C Flash Pt: 93°C |
MDL Number:
MFCD00005724 CAS Number: 1640-39-7 EINECS: 216-685-6 |
Risultati del test di specificazione
Appearance | Light yellow to Brown clear liquid to slightly cloudy liquid |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...