Stampa…

N-Methylnicotinamide ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(Methylcarbamoyl)pyridine

M0374-10G M0374-25G
TCIAM0374-10GEA 56 EUR
TCIAM0374-10G TCIAM0374-25G
N-Methylnicotinamide ≥98.0% (by GC, titration analysis)
N-Methylnicotinamide
Formula: C₇H₈N₂O
Peso molecolare: 136,1512 g/mol
MDL Number: MFCD00006384
CAS Number: 114-33-0
EINECS: 204-046-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa97eeb779872c29a275982313f5392a474b9028&vid=aa97eeb779872c29a275982313f5392a474b9028&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR