Stampa…

6-Aminonicotinamide 99%

Fornitore: Thermo Scientific
Pericolo

Synonyms: 6-Aminopyridine-3-carboxamide

L06692.03 L06692.06
L06692.03EA 61.3 EUR
L06692.03 L06692.06
6-Aminonicotinamide 99%
6-Aminonicotinamide

Purity: 99% CAS No.: 329-89-5 Molecular Formula: C6H7N3O Formula Weight: 137.14 Melting Point: 246-248° Beilstein Reference: 116042

Formula: C₆H₇N₃O
Peso molecolare: 137,14 g/mol
Punto di fusione: 246…248 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006327
CAS Number: 329-89-5
EINECS: 206-349-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR