Stampa…

4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: 5,5'-(Perfluoropropane-2,2-diyl)bis(isobenzofuran-1,3-dione), 2,2-Bis(1,3-dihydro-1,3-dioxobenzo[c]furan-5-yl)hexafluoropropane

H0771-25G H0771-5G H0771-100G
TCIAH0771-25GEA 124 EUR
TCIAH0771-25G TCIAH0771-5G TCIAH0771-100G
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥98.0% (by GC, titration analysis)
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
Formula: C₈H₄O₃
Peso molecolare: 444.25 g/mol
MDL Number: MFCD00039143
CAS Number: 1107-00-2
EINECS: 214-170-0
UN: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=595f8f57987d13c80922138e5c1322fc4315a197&vid=595f8f57987d13c80922138e5c1322fc4315a197&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 98.0 %
Melting point 243.0 to 249.0 deg-C
Solubility in hot Acetonitrile almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR