Stampa…

3,4,5-Trimethoxybenzoic anhydride ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

T1093-25G T1093-5G
TCIAT1093-25GEA 288 EUR
TCIAT1093-25G TCIAT1093-5G
3,4,5-Trimethoxybenzoic anhydride ≥95.0% (by titrimetric analysis)
3,4,5-Trimethoxybenzoic anhydride
Formula: C₂₀H₂₂O₉
Peso molecolare: 406,383 g/mol
MDL Number: MFCD00017155
CAS Number: 1719-88-6
EINECS: 217-010-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a164d180281f516daf5d4ecc0f68357ed702747c&vid=a164d180281f516daf5d4ecc0f68357ed702747c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Morpholine Method) min. 95.0 %
Melting point 156.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR