4-tert-Butylbenzoic anhydride 95%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 95%
CAS No.: 22201-45-2
Molecular Formula: C22H26O3
Formula Weight: 338.45
Melting Point: 75-81°
MDL No.: MFCD09757530
Formula:
C₂₂H₂₆O₃ Punto di fusione: 75…81 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09757530 CAS Number: 22201-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...