Stampa…

2-Amino-2'-chloro-5-nitrobenzophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Clonazepam Related Compound B, (2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone

A1754-25G A1754-5G
TCIAA1754-25GEA 196 EUR
TCIAA1754-25G TCIAA1754-5G
2-Amino-2'-chloro-5-nitrobenzophenone ≥98.0% (by GC)
2-Amino-2'-chloro-5-nitrobenzophenone
Formula: C₁₃H₉ClN₂O₃
Peso molecolare: 276,68 g/mol
MDL Number: MFCD00792453
CAS Number: 2011-66-7
EINECS: 217-929-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2a5c7e443d048e20ab8f9de3882e2ed4fb0d4bc&vid=f2a5c7e443d048e20ab8f9de3882e2ed4fb0d4bc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR