2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Attenzione
|
Synonyms:
5-Chloro-4-methylaniline-2-sulphonic acid Sodium Salt, Sodium 2-amino-4-chloro-5-methylbenzenesulphonate, Aminochlorotoluenesulphonicacidsodiumsalt, 5-Chloro-p-toluidine-2-sulphonic acid Sodium Salt, Sodium 2-Amino-4-chloro-5-methylbenzenesulphonate
Formula:
C₇H₇ClNNaO₃S |
MDL Number:
MFCD00070551 CAS Number: 6627-59-4 EINECS: 229-596-2 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...