2'-Chloro-4'-(4-chlorophenoxy)acetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-[2-Chloro-4-(4-chlorophenoxy)phenyl]ethan-1-one, 4-Acetyl-3,4'-dichlorodiphenyl ether
C2219-25G
C2219-5G
TCIAC2219-25GEA
200
EUR
TCIAC2219-25G
TCIAC2219-5G
2'-Chloro-4'-(4-chlorophenoxy)acetophenone ≥98.0% (by GC)
2'-Chloro-4'-(4-chlorophenoxy)acetophenone
Formula:
C₁₄H₁₀Cl₂O₂ Peso molecolare: 281,1340 g/mol |
MDL Number:
MFCD00140226 CAS Number: 119851-28-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...