Stampa…

4'-Amino-3',5'-dichloroacetophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4′-Amino-3′,5′-dichloroacetophenone

A1967-25G A1967-5G
TCIAA1967-25GEA 76 EUR
TCIAA1967-25G TCIAA1967-5G
4'-Amino-3',5'-dichloroacetophenone ≥98.0% (by GC)
4'-Amino-3',5'-dichloroacetophenone
Formula: C₈H₇Cl₂NO
Peso molecolare: 204,0500 g/mol
MDL Number: MFCD00238535
CAS Number: 37148-48-4
EINECS: 253-368-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8f14bee008c9d285297d52fd1d937ff668310c5&vid=a8f14bee008c9d285297d52fd1d937ff668310c5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 162.0 to 166.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR