Stampa…

3,3',4,5'-Tetramethoxypiceatannol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (E)-4-(3,5-Dimethoxystyryl)-1,2-dimethoxybenzene

T2842-1G T2842-5G
TCIAT2842-1GEA 97 EUR
TCIAT2842-1G TCIAT2842-5G
3,3',4,5'-Tetramethoxypiceatannol ≥98.0% (by GC)
3,3',4,5'-Tetramethoxypiceatannol
Formula: C₁₈H₂₀O₄
Peso molecolare: 300,3490 g/mol
MDL Number: MFCD09033675
CAS Number: 83088-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f340bc9b8a03c4e6974bd28881450a365b8de57&vid=7f340bc9b8a03c4e6974bd28881450a365b8de57&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR