Stampa…

4-Hydroxydiphenylamine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

A0470-25G A0470-500G
TCIAA0470-25GEA 36 EUR
TCIAA0470-25G TCIAA0470-500G
4-Hydroxydiphenylamine ≥98.0% (by HPLC, titration analysis)
4-Hydroxydiphenylamine
Formula: C₁₂H₁₁NO
Punto di ebollizione: 165°C
MDL Number: MFCD00020142
CAS Number: 122-37-2
EINECS: 204-538-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb23d50248a2513e7f4293fca9a77ed8d5faadad&vid=fb23d50248a2513e7f4293fca9a77ed8d5faadad&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 70.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR