4-(Isopropylamino)diphenylamine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₅H₁₈N₂ Punto di ebollizione: 180°C Flash Pt: 198°C |
MDL Number:
MFCD00026348 CAS Number: 101-72-4 EINECS: 202-969-7 UN: 3077 ADR: 9,III |
|
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 77.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...