Stampa…

5-Chloro-2-nitrodiphenylamine ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 5-Chloro-2-nitro-N-phenylaniline

C1785-25G C1785-5G
TCIAC1785-25GEA 79 EUR
TCIAC1785-25G TCIAC1785-5G
5-Chloro-2-nitrodiphenylamine ≥98.0% (by GC)
5-Chloro-2-nitrodiphenylamine
Formula: C₁₂H₉ClN₂O₂
MDL Number: MFCD00007287
CAS Number: 25781-92-4
EINECS: 247-261-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03b4e61c0fee6df8ab49af81825fd32bef4f4ce8&vid=03b4e61c0fee6df8ab49af81825fd32bef4f4ce8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 116.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR