Stampa…

2-Chloro-4-fluoro-5-nitrophenol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C2616-5G C2616-25G
TCIAC2616-5GEA 241 EUR
TCIAC2616-5G TCIAC2616-25G
2-Chloro-4-fluoro-5-nitrophenol ≥98.0% (by GC, titration analysis)
2-Chloro-4-fluoro-5-nitrophenol
Formula: C₆H₃ClFNO₃
Peso molecolare: 191,5400 g/mol
MDL Number: MFCD02670258
CAS Number: 84478-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abce862822967d011490d27116cea6263d471c2e&vid=abce862822967d011490d27116cea6263d471c2e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR