Stampa…
Hesperetin
Fornitore: Cayman Chemical
Synonyms: 3',5,7-Trihydroxy-4'-methoxyflavanone, 4'-Methoxy-3',5,7-trihydroxyflavanone, (S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (±)-Hesperetin
10006084-25 10006084-50 10006084-100 10006084-500
CAYM10006084-25EA 110 EUR
CAYM10006084-25 CAYM10006084-50 CAYM10006084-100 CAYM10006084-500
Hesperetin
Hesperetin
Formula: C₁₆H₁₄O₆
Peso molecolare: 302,30 g/mol
MDL Number: MFCD00075646
CAS Number: 520-33-2
EINECS: 208-290-2