Stampa…

2,3-Dimethylmaleic anhydride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,4-Dimethylfuran-2,5-dione

D3473-25G D3473-5G
TCIAD3473-25GEA 142 EUR
TCIAD3473-25G TCIAD3473-5G
2,3-Dimethylmaleic anhydride ≥98.0% (by GC, titration analysis)
2,3-Dimethylmaleic anhydride
Formula: C₆H₆O₃
Peso molecolare: 126,11 g/mol
Punto di ebollizione: 119°C
MDL Number: MFCD00005523
CAS Number: 766-39-2
EINECS: 212-165-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5946f01f27d59655604a170e65df04953bfadeef&vid=5946f01f27d59655604a170e65df04953bfadeef&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Methanolysis Method) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR